Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTTTCTTTGGTCCATTAAGGTCCA[C/T]GTTTGAAGTATCAGTAGGAATGCTT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 610510 MIM: 607533 | ||||||||||||||||||||
Literature Links: |
BORA PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
BORA - bora, aurora kinase A activator | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
DIS3 - DIS3 homolog, exosome endoribonuclease and 3'-5' exoribonuclease | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001128226.2 | 3126 | Missense Mutation | ATG,GTG | M,V 914 | NP_001121698.1 | |
NM_001322348.1 | 3126 | Missense Mutation | ATG,GTG | M,V 821 | NP_001309277.1 | |
NM_001322349.1 | 3126 | Missense Mutation | ATG,GTG | M,V 782 | NP_001309278.1 | |
NM_014953.4 | 3126 | Missense Mutation | ATG,GTG | M,V 944 | NP_055768.3 |