Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACTGATCCATAATAATAAAAATGTC[C/T]CGAAAAATTTCAAAGGAGTCAAAAA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 607533 MIM: 607532 | ||||||||||||||||||||
Literature Links: |
DIS3 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
DIS3 - DIS3 homolog, exosome endoribonuclease and 3'-5' exoribonuclease | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PIBF1 - progesterone immunomodulatory binding factor 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006346.2 | 339 | Silent Mutation | TCC,TCT | S,S 2 | NP_006337.2 | |
XM_011534881.2 | 339 | Silent Mutation | TCC,TCT | S,S 2 | XP_011533183.1 | |
XM_011534882.2 | 339 | Silent Mutation | TCC,TCT | S,S 2 | XP_011533184.1 | |
XM_011534884.2 | 339 | Silent Mutation | TCC,TCT | S,S 2 | XP_011533186.1 | |
XM_011534885.2 | 339 | Intron | XP_011533187.1 | |||
XM_011534886.2 | 339 | Silent Mutation | TCC,TCT | S,S 2 | XP_011533188.1 | |
XM_017020350.1 | 339 | Intron | XP_016875839.1 | |||
XM_017020351.1 | 339 | Silent Mutation | TCC,TCT | S,S 2 | XP_016875840.1 | |
XM_017020352.1 | 339 | UTR 5 | XP_016875841.1 | |||
XM_017020353.1 | 339 | UTR 5 | XP_016875842.1 | |||
XM_017020354.1 | 339 | UTR 5 | XP_016875843.1 |