Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGCGTAGCCGGTGCTGGCTTCTTT[C/T]TTGCATTTTTGGTTTTGCATTCGCG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
NHLRC3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
NHLRC3 - NHL repeat containing 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001012754.3 | 372 | Missense Mutation | CTT,TTT | L,F 15 | NP_001012772.1 | |
NM_001017370.2 | 372 | Missense Mutation | CTT,TTT | L,F 15 | NP_001017370.1 |
PROSER1 - proline and serine rich 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_025138.4 | 372 | Intron | NP_079414.3 | |||
XM_005266545.3 | 372 | Intron | XP_005266602.1 | |||
XM_011535239.2 | 372 | Intron | XP_011533541.1 |