Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TAGAAAAATCTGAGTTTTGAACTGA[C/T]CAACTTCAAGTCTTTTAAAATCTTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604742 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
DCLK1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DCLK1 - doublecortin like kinase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001195415.1 | 5654 | UTR 3 | NP_001182344.1 | |||
NM_001195416.1 | 5654 | UTR 3 | NP_001182345.1 | |||
NM_001195430.1 | 5654 | Intron | NP_001182359.1 | |||
NM_004734.4 | 5654 | UTR 3 | NP_004725.1 | |||
XM_005266592.2 | 5654 | UTR 3 | XP_005266649.1 | |||
XM_006719893.2 | 5654 | UTR 3 | XP_006719956.1 | |||
XM_017020847.1 | 5654 | UTR 3 | XP_016876336.1 | |||
XM_017020848.1 | 5654 | Intron | XP_016876337.1 |