Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGAAGTGCACCTGCCACCGGCCTGG[A/C]AAGAAGGAGACGTGGCTCTTCTCCC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 609401 | ||||||||||||||||||||
Literature Links: |
HS6ST3 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
HS6ST3 - heparan sulfate 6-O-sulfotransferase 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_153456.3 | 799 | Silent Mutation | GGA,GGC | G,G 190 | NP_703157.2 | |
XM_011521076.2 | 799 | Silent Mutation | GGA,GGC | G,G 190 | XP_011519378.1 | |
XM_017020543.1 | 799 | Silent Mutation | GGA,GGC | G,G 190 | XP_016876032.1 |