Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CATTTGTAGCATTGATTTCCCGGAT[C/T]TGGTAACATGGCAAAAGATGTAAGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 189902 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
TFDP1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
TFDP1 - transcription factor Dp-1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_007111.4 | Intron | NP_009042.1 | ||||
XM_005268327.1 | Intron | XP_005268384.1 | ||||
XM_005268328.2 | Intron | XP_005268385.1 | ||||
XM_005268331.1 | Intron | XP_005268388.1 | ||||
XM_011537518.2 | Intron | XP_011535820.1 | ||||
XM_011537519.1 | Intron | XP_011535821.1 | ||||
XM_011537520.2 | Intron | XP_011535822.1 | ||||
XM_017020717.1 | Intron | XP_016876206.1 | ||||
XM_017020718.1 | Intron | XP_016876207.1 | ||||
XM_017020719.1 | Intron | XP_016876208.1 | ||||
XM_017020720.1 | Intron | XP_016876209.1 | ||||
XM_017020721.1 | Intron | XP_016876210.1 | ||||
XM_017020722.1 | Intron | XP_016876211.1 |