Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGCCTTCTACCCTTTGTCTACAGG[A/C]ATTCTGGAATTTATCAGTATAGCAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 600921 | ||||||||||||||||||||
Literature Links: |
FGF9 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
FGF9 - fibroblast growth factor 9 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002010.2 | 1116 | Silent Mutation | GGA,GGC | G,G 93 | NP_002001.1 | |
XM_011534996.2 | 1116 | Silent Mutation | GGA,GGC | G,G 43 | XP_011533298.1 |