Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGCTCACCTTTTCATTCACTCCGTC[A/G]CTATTGTCTTTAGAAGCCTCTTCAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 182135 | ||||||||||||||||||||
Literature Links: |
HTR2A PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
HTR2A - 5-hydroxytryptamine receptor 2A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000621.4 | 1374 | Silent Mutation | AGC,AGT | S,S 461 | NP_000612.1 | |
NM_001165947.2 | 1374 | Silent Mutation | AGC,AGT | S,S 377 | NP_001159419.1 |