Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATTTGATTTGCGAGAGGAGAAAATA[A/C]ATTTCAAGGTTTTTTTCTTTTTTTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 163905 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
HMGB1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
HMGB1 - high mobility group box 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
USPL1 - ubiquitin specific peptidase like 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001321532.1 | Intron | NP_001308461.1 | ||||
NM_001321533.1 | Intron | NP_001308462.1 | ||||
NM_001321534.1 | Intron | NP_001308463.1 | ||||
NM_005800.4 | Intron | NP_005791.3 | ||||
XM_005266213.3 | Intron | XP_005266270.1 | ||||
XM_006719751.3 | Intron | XP_006719814.1 | ||||
XM_017020314.1 | Intron | XP_016875803.1 | ||||
XM_017020315.1 | Intron | XP_016875804.1 | ||||
XM_017020316.1 | Intron | XP_016875805.1 |