Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATGTGAAGGCTCGGAGCTGCGGAGA[C/G]GTCCGCCAGGCGTACGGTGCCAAGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 604404 | ||||||||||||||||||||
Literature Links: |
GPC6 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
GPC6 - glypican 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005708.3 | 728 | Missense Mutation | GAC,GAG | D,E 32 | NP_005699.1 | |
XM_011521044.2 | 728 | Intron | XP_011519346.1 | |||
XM_017020298.1 | 728 | Intron | XP_016875787.1 | |||
XM_017020299.1 | 728 | Intron | XP_016875788.1 | |||
XM_017020300.1 | 728 | Intron | XP_016875789.1 | |||
XM_017020301.1 | 728 | Intron | XP_016875790.1 | |||
XM_017020302.1 | 728 | Intron | XP_016875791.1 |