Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTCAGTGTGCCTTCCACAGTTGCCA[C/T]GGCAACTAGCCTATCACACACAGCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 182135 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
HTR2A PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
HTR2A - 5-hydroxytryptamine receptor 2A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000621.4 | 1421 | UTR 3 | NP_000612.1 | |||
NM_001165947.2 | 1421 | UTR 3 | NP_001159419.1 |