Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGTTACCTTTTCCCCTCTTTGTGTG[A/G]TAAATTGCTTGGTTTTAGTTACTGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604098 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SUGT1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SUGT1 - SGT1 homolog, MIS12 kinetochore complex assembly cochaperone | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001130912.2 | Intron | NP_001124384.1 | ||||
NM_001320831.1 | Intron | NP_001307760.1 | ||||
NM_006704.4 | Intron | NP_006695.1 | ||||
XM_011534891.2 | Intron | XP_011533193.1 | ||||
XM_011534893.2 | Intron | XP_011533195.1 | ||||
XM_017020365.1 | Intron | XP_016875854.1 | ||||
XM_017020366.1 | Intron | XP_016875855.1 | ||||
XM_017020367.1 | Intron | XP_016875856.1 | ||||
XM_017020368.1 | Intron | XP_016875857.1 |