Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGTGGCGAGCCTCCACGTTCCGCCA[A/C]CGTACAGATCAACGTGAAGGTGATT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611760 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PCDH17 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PCDH17 - protocadherin 17 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001040429.2 | 3120 | Missense Mutation | AAC,ACC | N,T 226 | NP_001035519.1 | |
XM_005266357.2 | 3120 | Missense Mutation | AAC,ACC | N,T 226 | XP_005266414.1 | |
XM_005266358.2 | 3120 | Missense Mutation | AAC,ACC | N,T 226 | XP_005266415.1 | |
XM_017020547.1 | 3120 | Missense Mutation | AAC,ACC | N,T 226 | XP_016876036.1 |