Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGTAGCTGAAGCACAAGAAAATAAA[C/G]TTGGAAAGAAAAAACCTAAGGCCAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610510 MIM: 607533 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
BORA PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
BORA - bora, aurora kinase A activator | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001286746.1 | 5808 | Intron | NP_001273675.1 | |||
NM_001286747.1 | 5808 | Intron | NP_001273676.1 | |||
NM_024808.3 | 5808 | Intron | NP_079084.3 | |||
XM_006719868.3 | 5808 | Intron | XP_006719931.2 |
DIS3 - DIS3 homolog, exosome endoribonuclease and 3'-5' exoribonuclease | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001128226.2 | 5808 | UTR 3 | NP_001121698.1 | |||
NM_001322348.1 | 5808 | UTR 3 | NP_001309277.1 | |||
NM_001322349.1 | 5808 | UTR 3 | NP_001309278.1 | |||
NM_014953.4 | 5808 | UTR 3 | NP_055768.3 |