Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTTTGGAAGCTGCTCAGAAACTCA[A/G]TGAGTTTGGGCTGATTTTTTAACAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 612175 | ||||||||||||||||||||
Literature Links: |
CAB39L PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CAB39L - calcium binding protein 39 like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001079670.2 | 1833 | Missense Mutation | ACT,ATT | T,I 302 | NP_001073138.1 | |
NM_001287337.1 | 1833 | Missense Mutation | ACT,ATT | T,I 302 | NP_001274266.1 | |
NM_001287338.1 | 1833 | Missense Mutation | ACT,ATT | T,I 302 | NP_001274267.1 | |
NM_001287339.1 | 1833 | Missense Mutation | ACT,ATT | T,I 302 | NP_001274268.1 | |
NM_030925.3 | 1833 | Missense Mutation | ACT,ATT | T,I 302 | NP_112187.2 | |
XM_011535254.2 | 1833 | Missense Mutation | ACT,ATT | T,I 302 | XP_011533556.1 | |
XM_011535255.2 | 1833 | Missense Mutation | ACT,ATT | T,I 302 | XP_011533557.1 | |
XM_011535256.2 | 1833 | Missense Mutation | ACT,ATT | T,I 302 | XP_011533558.1 | |
XM_017020785.1 | 1833 | Missense Mutation | ACT,ATT | T,I 302 | XP_016876274.1 | |
XM_017020786.1 | 1833 | Missense Mutation | ACT,ATT | T,I 302 | XP_016876275.1 | |
XM_017020787.1 | 1833 | Missense Mutation | ACT,ATT | T,I 302 | XP_016876276.1 | |
XM_017020788.1 | 1833 | Missense Mutation | ACT,ATT | T,I 302 | XP_016876277.1 |