Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCAGGCAAATCTGTTGCTGTGACTT[G/T]AGCTCCTAAGAGAAAAAGAAAACAA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 615259 MIM: 190470 | ||||||||||||||||||||
Literature Links: |
METTL21C PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
METTL21C - methyltransferase like 21C | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001010977.2 | 429 | Missense Mutation | AAA,CAA | K,Q 136 | NP_001010977.1 | |
XM_017020407.1 | 429 | Missense Mutation | AAA,CAA | K,Q 136 | XP_016875896.1 | |
XM_017020408.1 | 429 | Missense Mutation | AAA,CAA | K,Q 136 | XP_016875897.1 |
TPP2 - tripeptidyl peptidase 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |