Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTGTTTTCTTTATTCCTTCTTTCCA[A/G]GGCACTTTAGGTCTCCATCCTAAGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616146 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
TGDS PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
TGDS - TDP-glucose 4,6-dehydratase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001304430.1 | 1172 | Silent Mutation | CCC,CCT | P,P 287 | NP_001291359.1 | |
NM_014305.3 | 1172 | Silent Mutation | CCC,CCT | P,P 319 | NP_055120.1 | |
XM_011521065.2 | 1172 | Silent Mutation | CCC,CCT | P,P 287 | XP_011519367.1 | |
XM_011521066.2 | 1172 | Silent Mutation | CCC,CCT | P,P 287 | XP_011519368.1 |