Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTCCTGGGGGTCGTCAATGACGAA[C/T]GTCGAATACTCCTCCTGCTCCGGGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605182 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC100507747 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LOC100507747 - uncharacterized LOC100507747 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RASA3 - RAS p21 protein activator 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001320821.1 | 2560 | Silent Mutation | NP_001307750.1 | |||
NM_001320822.1 | 2560 | Silent Mutation | NP_001307751.1 | |||
NM_007368.3 | 2560 | Silent Mutation | NP_031394.2 | |||
XM_011534841.2 | 2560 | Silent Mutation | XP_011533143.1 | |||
XM_011534842.2 | 2560 | Silent Mutation | XP_011533144.1 | |||
XM_017020438.1 | 2560 | Silent Mutation | XP_016875927.1 | |||
XM_017020439.1 | 2560 | Silent Mutation | XP_016875928.1 |