Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACTTGTTCAATATGTGTCTACAGCA[A/G]CTGTTTGAAGTAAAAGTTTTCAAGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 610326 | ||||||||||||||||||||
Literature Links: |
RNASEH2B PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
RNASEH2B - ribonuclease H2 subunit B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001142279.2 | 498 | Silent Mutation | CAA,CAG | Q,Q 59 | NP_001135751.1 | |
NM_024570.3 | 498 | Silent Mutation | CAA,CAG | Q,Q 59 | NP_078846.2 | |
XM_006719867.3 | 498 | Silent Mutation | CAA,CAG | Q,Q 53 | XP_006719930.1 | |
XM_011535229.1 | 498 | Silent Mutation | CAA,CAG | Q,Q 59 | XP_011533531.1 | |
XM_011535230.2 | 498 | Silent Mutation | CAA,CAG | Q,Q 59 | XP_011533532.1 | |
XM_011535231.2 | 498 | Silent Mutation | CAA,CAG | Q,Q 59 | XP_011533533.1 | |
XM_011535233.2 | 498 | UTR 5 | XP_011533535.1 | |||
XM_017020747.1 | 498 | Silent Mutation | CAA,CAG | Q,Q 59 | XP_016876236.1 |