Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTTGGAAATGCTCCAGTTTTTCAGG[A/G]AACAGGTGGGCACCTCGACTCTGAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 616821 | ||||||||||||||||||||
Literature Links: |
THSD1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
THSD1 - thrombospondin type 1 domain containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_018676.3 | 2660 | Silent Mutation | TTC,TTT | F,F 705 | NP_061146.1 | |
NM_199263.2 | 2660 | Silent Mutation | TTC,TTT | F,F 652 | NP_954872.1 |