Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTCACGAGATTTGGAGCTCGCGGGA[A/G]AACTTGTCTCTGCGTTGTGGGGAGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600405 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
RFC3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
RFC3 - replication factor C subunit 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002915.3 | 52 | UTR 5 | NP_002906.1 | |||
NM_181558.2 | 52 | UTR 5 | NP_853536.2 | |||
XM_011535172.2 | 52 | UTR 5 | XP_011533474.1 | |||
XM_011535173.2 | 52 | UTR 5 | XP_011533475.1 | |||
XM_011535174.2 | 52 | UTR 5 | XP_011533476.1 | |||
XM_011535175.1 | 52 | UTR 5 | XP_011533477.1 | |||
XM_017020680.1 | 52 | UTR 5 | XP_016876169.1 | |||
XM_017020681.1 | 52 | UTR 5 | XP_016876170.1 | |||
XM_017020682.1 | 52 | UTR 5 | XP_016876171.1 | |||
XM_017020683.1 | 52 | UTR 5 | XP_016876172.1 |