Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAGCCCAGGGTAAGTCCTGTTCTCA[A/G]AAGCCAACCCAGCTGCTTGTCTGGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610620 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ADPRHL1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ADPRHL1 - ADP-ribosylhydrolase like 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
DCUN1D2 - defective in cullin neddylation 1 domain containing 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001014283.1 | 6341 | UTR 3 | NP_001014305.1 | |||
XM_017020643.1 | 6341 | UTR 3 | XP_016876132.1 | |||
XM_017020644.1 | 6341 | UTR 3 | XP_016876133.1 | |||
XM_017020645.1 | 6341 | UTR 3 | XP_016876134.1 | |||
XM_017020646.1 | 6341 | UTR 3 | XP_016876135.1 | |||
XM_017020647.1 | 6341 | UTR 3 | XP_016876136.1 | |||
XM_017020648.1 | 6341 | UTR 3 | XP_016876137.1 | |||
XM_017020649.1 | 6341 | UTR 3 | XP_016876138.1 |