Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATTATTTGAAAAATACTTGAAGGAC[A/G]AAAGTGGCTTCAAAGATCCTTCCAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 603461 | ||||||||||||||||||||
Literature Links: |
CDC16 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CDC16 - cell division cycle 16 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
XM_005266206.1 | 516 | Missense Mutation | AAA,GAA | K,E 111 | XP_005266263.1 | |
XM_006719992.1 | 516 | Missense Mutation | AAA,GAA | K,E 110 | XP_006720055.1 | |
XM_006719993.1 | 516 | Missense Mutation | AAA,GAA | K,E 17 | XP_006720056.1 | |
XM_011534851.1 | 516 | Missense Mutation | AAA,GAA | K,E 111 | XP_011533153.1 | |
XM_011534853.1 | 516 | Missense Mutation | AAA,GAA | K,E 111 | XP_011533155.1 | |
XM_017020830.1 | 516 | Missense Mutation | AAA,GAA | K,E 111 | XP_016876319.1 | |
XM_017020831.1 | 516 | Missense Mutation | AAA,GAA | K,E 111 | XP_016876320.1 | |
XM_017020832.1 | 516 | Missense Mutation | AAA,GAA | K,E 17 | XP_016876321.1 | |
XM_017020833.1 | 516 | Missense Mutation | AAA,GAA | K,E 17 | XP_016876322.1 |
MIR548AR - microRNA 548ar | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |