Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTGTACTTTGGAAAGATTGCTCTA[A/G]TTGTATTATGAATTCCCAACCTTAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616443 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZMYM5 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ZMYM5 - zinc finger MYM-type containing 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001039649.2 | Intron | NP_001034738.1 | ||||
NM_001039650.2 | Intron | NP_001034739.1 | ||||
NM_001142684.1 | Intron | NP_001136156.1 | ||||
XM_005266593.3 | Intron | XP_005266650.1 | ||||
XM_005266594.2 | Intron | XP_005266651.1 | ||||
XM_006719894.2 | Intron | XP_006719957.1 | ||||
XM_011535309.1 | Intron | XP_011533611.1 | ||||
XM_011535310.1 | Intron | XP_011533612.1 | ||||
XM_017020849.1 | Intron | XP_016876338.1 | ||||
XM_017020850.1 | Intron | XP_016876339.1 | ||||
XM_017020851.1 | Intron | XP_016876340.1 |