Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGGGCATCTCCTGGACGATGTACAC[C/T]GGGTGCCCGTAGTCCCCGCTGACCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600527 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
EFNB2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
EFNB2 - ephrin B2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004093.3 | 1018 | Silent Mutation | CCA,CCG | P,P 314 | NP_004084.1 | |
XM_017020406.1 | 1018 | Silent Mutation | CCA,CCG | P,P 316 | XP_016875895.1 |