Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCGGCGCGGATGGCACGCCGGCGGA[A/G]CCAGCGAGTCTGCGCGAGCGGTCCG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610041 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
NDFIP2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
NDFIP2 - Nedd4 family interacting protein 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001161407.1 | 499 | Missense Mutation | AAC,AGC | N,S 6 | NP_001154879.1 | |
NM_019080.2 | 499 | Missense Mutation | AAC,AGC | N,S 6 | NP_061953.2 | |
XM_006719840.3 | 499 | Missense Mutation | AAC,AGC | N,S 6 | XP_006719903.1 |
NDFIP2-AS1 - NDFIP2 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |