Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTTGTTCTTTTTTTTCAGGTGCAG[C/T]GTGGTGACTTTCCTCATCTGTTAGT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 600405 | ||||||||||||||||||||
Literature Links: |
RFC3 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
RFC3 - replication factor C subunit 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002915.3 | 224 | Missense Mutation | CGT,TGT | R,C 32 | NP_002906.1 | |
NM_181558.2 | 224 | Missense Mutation | CGT,TGT | R,C 32 | NP_853536.2 | |
XM_011535172.2 | 224 | Missense Mutation | CGT,TGT | R,C 32 | XP_011533474.1 | |
XM_011535173.2 | 224 | Missense Mutation | CGT,TGT | R,C 32 | XP_011533475.1 | |
XM_011535174.2 | 224 | Missense Mutation | CGT,TGT | R,C 32 | XP_011533476.1 | |
XM_011535175.1 | 224 | Missense Mutation | CGT,TGT | R,C 32 | XP_011533477.1 | |
XM_017020680.1 | 224 | Missense Mutation | CGT,TGT | R,C 32 | XP_016876169.1 | |
XM_017020681.1 | 224 | Missense Mutation | CGT,TGT | R,C 32 | XP_016876170.1 | |
XM_017020682.1 | 224 | Missense Mutation | CGT,TGT | R,C 32 | XP_016876171.1 | |
XM_017020683.1 | 224 | Missense Mutation | CGT,TGT | R,C 32 | XP_016876172.1 |