Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TAAGAACAGGTTTTTAATATTGTGT[A/G]TATTTTTAAAAGATGTCTTATTATT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603295 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SMAD9 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SMAD9 - SMAD family member 9 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001127217.2 | 2854 | UTR 3 | NP_001120689.1 | |||
NM_005905.5 | 2854 | UTR 3 | NP_005896.1 | |||
XM_005266401.3 | 2854 | Intron | XP_005266458.1 | |||
XM_005266403.3 | 2854 | Intron | XP_005266460.1 | |||
XM_005266404.3 | 2854 | Intron | XP_005266461.1 | |||
XM_006719827.3 | 2854 | Intron | XP_006719890.1 |