Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTCAGCAGACCGTAATTCTCCTTA[C/G]TTCCTGATGCCGTGGGGTCAAGCTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
TMTC4 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
TMTC4 - transmembrane and tetratricopeptide repeat containing 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001079669.2 | 2478 | Missense Mutation | ACT,AGT | T,S 722 | NP_001073137.1 | |
NM_001286453.1 | 2478 | Missense Mutation | ACT,AGT | T,S 611 | NP_001273382.1 | |
NM_032813.3 | 2478 | Missense Mutation | ACT,AGT | T,S 741 | NP_116202.2 | |
XM_011521121.2 | 2478 | Missense Mutation | ACT,AGT | T,S 799 | XP_011519423.1 | |
XM_011521122.2 | 2478 | Missense Mutation | ACT,AGT | T,S 799 | XP_011519424.1 | |
XM_011521123.2 | 2478 | Missense Mutation | ACT,AGT | T,S 780 | XP_011519425.1 | |
XM_011521124.2 | 2478 | Missense Mutation | ACT,AGT | T,S 780 | XP_011519426.1 | |
XM_011521125.1 | 2478 | Intron | XP_011519427.1 | |||
XM_017020796.1 | 2478 | Missense Mutation | ACT,AGT | T,S 722 | XP_016876285.1 | |
XM_017020797.1 | 2478 | Missense Mutation | ACT,AGT | T,S 563 | XP_016876286.1 | |
XM_017020798.1 | 2478 | Missense Mutation | ACT,AGT | T,S 505 | XP_016876287.1 | |
XM_017020799.1 | 2478 | Missense Mutation | ACT,AGT | T,S 505 | XP_016876288.1 | |
XM_017020800.1 | 2478 | Intron | XP_016876289.1 |