Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TACTTGCCTCTTGCAATCGAAAAAC[A/G]ACGGGGATTTTCTTTTCTGTACAGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 133280 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ESD PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ESD - esterase D | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001984.1 | 997 | Silent Mutation | GTC,GTT | V,V 250 | NP_001975.1 | |
XM_005266278.2 | 997 | Silent Mutation | GTC,GTT | V,V 250 | XP_005266335.1 | |
XM_011534954.1 | 997 | Silent Mutation | GTC,GTT | V,V 250 | XP_011533256.1 | |
XM_017020414.1 | 997 | Silent Mutation | GTC,GTT | V,V 131 | XP_016875903.1 |