Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACCTGGCAATCTTCCTGCTGCAAGT[A/G]CATCTCCTGGTAAATGGCCATTCAC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 605718 | ||||||||||||||||||||
Literature Links: |
MED4 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
MED4 - mediator complex subunit 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001270629.1 | 680 | Missense Mutation | GCA,GTA | A,V 160 | NP_001257558.1 | |
NM_014166.3 | 680 | Missense Mutation | GCA,GTA | A,V 206 | NP_054885.1 | |
XM_017020549.1 | 680 | Missense Mutation | GCA,GTA | A,V 184 | XP_016876038.1 |
MED4-AS1 - MED4 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |