Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGGTCAGAACTGTGGGATTGATGAC[C/T]TCCCACAACTGGAGCAGCCGGGAAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602008 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
IPO5 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
IPO5 - importin 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002271.4 | 38 | UTR 5 | NP_002262.3 | |||
XM_005254052.3 | 38 | Intron | XP_005254109.1 | |||
XM_005254053.3 | 38 | Intron | XP_005254110.1 | |||
XM_011521087.2 | 38 | UTR 5 | XP_011519389.1 | |||
XM_011521088.2 | 38 | UTR 5 | XP_011519390.1 | |||
XM_011521089.2 | 38 | UTR 5 | XP_011519391.1 | |||
XM_011521090.2 | 38 | Intron | XP_011519392.1 | |||
XM_017020562.1 | 38 | UTR 5 | XP_016876051.1 |