Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTTGGCAAGAAACTGAGCTCCGGCC[A/G]AGAACCGTGATTCCAGGTAGGGCCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
SPACA7 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SPACA7 - sperm acrosome associated 7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_145248.4 | 122 | Silent Mutation | CCA,CCG | P,P 26 | NP_660291.2 | |
XM_011537466.1 | 122 | Silent Mutation | CCA,CCG | P,P 43 | XP_011535768.1 | |
XM_011537467.2 | 122 | Silent Mutation | CCA,CCG | P,P 43 | XP_011535769.1 | |
XM_011537468.2 | 122 | UTR 5 | XP_011535770.1 | |||
XM_011537471.2 | 122 | UTR 5 | XP_011535773.1 | |||
XM_011537472.2 | 122 | Silent Mutation | CCA,CCG | P,P 43 | XP_011535774.1 | |
XM_017020391.1 | 122 | Silent Mutation | CCA,CCG | P,P 43 | XP_016875880.1 | |
XM_017020392.1 | 122 | UTR 5 | XP_016875881.1 |