Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTGAGCTGGGAGAGTGGAGGAGAGC[A/C]CAAGACTGGATATGAAACAGATGCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606977 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
COG6 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
COG6 - component of oligomeric golgi complex 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001145079.1 | Intron | NP_001138551.1 | ||||
NM_020751.2 | Intron | NP_065802.1 | ||||
XM_011535168.1 | Intron | XP_011533470.1 |
MIR4305 - microRNA 4305 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |