Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GACCCTCTGCTTTGTCCTCCTGCTG[C/T]GCTGTTGGCAAGAAACTGAGCTCCG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
SPACA7 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
SPACA7 - sperm acrosome associated 7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_145248.4 | 93 | Missense Mutation | CGC,TGC | R,C 17 | NP_660291.2 | |
XM_011537466.1 | 93 | Missense Mutation | CGC,TGC | R,C 34 | XP_011535768.1 | |
XM_011537467.2 | 93 | Missense Mutation | CGC,TGC | R,C 34 | XP_011535769.1 | |
XM_011537468.2 | 93 | UTR 5 | XP_011535770.1 | |||
XM_011537471.2 | 93 | UTR 5 | XP_011535773.1 | |||
XM_011537472.2 | 93 | Missense Mutation | CGC,TGC | R,C 34 | XP_011535774.1 | |
XM_017020391.1 | 93 | Missense Mutation | CGC,TGC | R,C 34 | XP_016875880.1 | |
XM_017020392.1 | 93 | UTR 5 | XP_016875881.1 |