Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCAGAAAAGAAACAGGTTGGTGATG[A/G]ACAAAAGGAGGAAAGAACTGCATAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614392 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
TDRD3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
TDRD3 - tudor domain containing 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001146070.1 | Intron | NP_001139542.1 | ||||
NM_001146071.1 | Intron | NP_001139543.1 | ||||
NM_030794.2 | Intron | NP_110421.1 | ||||
XM_005266556.4 | Intron | XP_005266613.1 | ||||
XM_005266560.2 | Intron | XP_005266617.1 | ||||
XM_011535247.1 | Intron | XP_011533549.1 | ||||
XM_011535249.2 | Intron | XP_011533551.1 | ||||
XM_017020777.1 | Intron | XP_016876266.1 |