Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACATGGCATGATGACTCTGCAGGCC[A/G]AACGGCAGCCTTGATAATTATAAGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 605147 MIM: 604098 | ||||||||||||||||||||
Literature Links: |
LECT1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
LECT1 - leukocyte cell derived chemotaxin 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001011705.1 | 939 | Missense Mutation | TCG,TTG | S,L 314 | NP_001011705.1 | |
NM_007015.2 | 939 | Missense Mutation | TCG,TTG | S,L 315 | NP_008946.1 | |
XM_011534897.2 | 939 | Missense Mutation | TCG,TTG | S,L 346 | XP_011533199.1 | |
XM_011534898.2 | 939 | Missense Mutation | TCG,TTG | S,L 345 | XP_011533200.1 | |
XM_011534899.2 | 939 | Missense Mutation | TCG,TTG | S,L 308 | XP_011533201.1 | |
XM_011534900.2 | 939 | Missense Mutation | TCG,TTG | S,L 304 | XP_011533202.1 |
SUGT1 - SGT1 homolog, MIS12 kinetochore complex assembly cochaperone | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |