Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCTCAGAGCCAGCTGGAAGGACTAT[C/G]TGGAAGGCACAGATATCCTCGTGTA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 609351 | ||||||||||||||||||||
Literature Links: |
ARL11 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ARL11 - ADP ribosylation factor like GTPase 11 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_138450.5 | 476 | Silent Mutation | CTG,GTG | L,V 78 | NP_612459.1 | |
XM_005266253.3 | 476 | Missense Mutation | CTG,GTG | L,V 78 | XP_005266310.1 |