Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CACAGTACTCGGCCACACCATGGGG[C/T]GCCCACTGCACCTCGTCCTGCTCAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613872 MIM: 613878 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
F10 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
F10 - coagulation factor X | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000504.3 | 64 | Missense Mutation | CGC,TGC | R,C 3 | NP_000495.1 | |
NM_001312674.1 | 64 | Missense Mutation | CGC,TGC | R,C 3 | NP_001299603.1 | |
NM_001312675.1 | 64 | Missense Mutation | CGC,TGC | R,C 3 | NP_001299604.1 |
F10-AS1 - F10 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
F7 - coagulation factor VII | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |