Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGCAAAGAAGACAAAAACTCAAGGA[A/G]CATCTGTTGAGAAGAAAAACGCTTT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 611569 MIM: 610903 | ||||||||||||||||||||
Literature Links: |
CKAP2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CKAP2 - cytoskeleton associated protein 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001098525.2 | 293 | Silent Mutation | GAA,GAG | E,E 32 | NP_001091995.1 | |
NM_001286686.1 | 293 | UTR 5 | NP_001273615.1 | |||
NM_001286687.1 | 293 | Silent Mutation | GAA,GAG | E,E 32 | NP_001273616.1 | |
NM_018204.4 | 293 | Silent Mutation | GAA,GAG | E,E 32 | NP_060674.3 | |
XM_005266344.4 | 293 | UTR 5 | XP_005266401.1 | |||
XM_011535043.2 | 293 | UTR 5 | XP_011533345.1 |
VPS36 - vacuolar protein sorting 36 homolog | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |