Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGAGAGCAACCTCTTCAAACGCATC[C/T]TGGTGAGTGGCCGTAGGCCGCGCTG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 604601 | ||||||||||||||||||||
Literature Links: |
MTRF1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
NAA16 - N(alpha)-acetyltransferase 16, NatA auxiliary subunit | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001110798.1 | 352 | Silent Mutation | CTG,TTG | L,L 18 | NP_001104268.1 | |
NM_018527.3 | 352 | Silent Mutation | CTG,TTG | L,L 18 | NP_060997.2 | |
NM_024561.4 | 352 | Silent Mutation | CTG,TTG | L,L 18 | NP_078837.3 | |
XM_005266523.2 | 352 | Intron | XP_005266580.1 | |||
XM_006719866.2 | 352 | Silent Mutation | CTG,TTG | L,L 18 | XP_006719929.1 | |
XM_011535227.2 | 352 | Silent Mutation | CTG,TTG | L,L 18 | XP_011533529.1 | |
XM_011535228.1 | 352 | Silent Mutation | CTG,TTG | L,L 18 | XP_011533530.1 | |
XM_017020744.1 | 352 | Intron | XP_016876233.1 | |||
XM_017020745.1 | 352 | Silent Mutation | CTG,TTG | L,L 18 | XP_016876234.1 | |
XM_017020746.1 | 352 | Silent Mutation | CTG,TTG | L,L 18 | XP_016876235.1 |