Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGCGACTTTCAGAAACAATCACAT[A/G]AAGACTAAAGCATCTGTCAGAAAAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 604696 | ||||||||||||||||||||
Literature Links: |
AKAP11 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
AKAP11 - A-kinase anchoring protein 11 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_016248.3 | 371 | Missense Mutation | ATA,ATG | I,M 9 | NP_057332.1 | |
XM_005266247.1 | 371 | Missense Mutation | ATA,ATG | I,M 9 | XP_005266304.1 | |
XM_005266248.3 | 371 | Missense Mutation | ATA,ATG | I,M 9 | XP_005266305.1 | |
XM_005266249.3 | 371 | Missense Mutation | ATA,ATG | I,M 9 | XP_005266306.1 | |
XM_005266250.3 | 371 | Missense Mutation | ATA,ATG | I,M 9 | XP_005266307.1 | |
XM_011534903.2 | 371 | Missense Mutation | ATA,ATG | I,M 9 | XP_011533205.1 | |
XM_011534904.1 | 371 | Missense Mutation | ATA,ATG | I,M 9 | XP_011533206.1 | |
XM_011534905.1 | 371 | Missense Mutation | ATA,ATG | I,M 9 | XP_011533207.1 | |
XM_011534906.2 | 371 | Missense Mutation | ATA,ATG | I,M 9 | XP_011533208.1 | |
XM_017020381.1 | 371 | Missense Mutation | ATA,ATG | I,M 9 | XP_016875870.1 | |
XM_017020382.1 | 371 | Missense Mutation | ATA,ATG | I,M 9 | XP_016875871.1 | |
XM_017020383.1 | 371 | Missense Mutation | ATA,ATG | I,M 9 | XP_016875872.1 |