Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGGGATGTTGAGGAGCTTCGCTGC[C/T]ACCAGGGGGTTGCTGTAGTGGGGCT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 137217 MIM: 180381 MIM: 189902 | ||||||||||||||||||||
Literature Links: |
ATP4B PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ATP4B - ATPase H+/K+ transporting beta subunit | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000705.3 | 792 | Silent Mutation | GTA,GTG | V,V 246 | NP_000696.1 |
GRK1 - G protein-coupled receptor kinase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002929.2 | 792 | Intron | NP_002920.1 | |||
XM_017020684.1 | 792 | Intron | XP_016876173.1 |
TFDP1 - transcription factor Dp-1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |