Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCAGTCAGGCCCCGGCGCAGGGTGT[C/T]GCATGCTTCCAGCAGGGGCTGCCTT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 612800 MIM: 615910 | ||||||||||||||||||||
Literature Links: |
CARS2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CARS2 - cysteinyl-tRNA synthetase 2, mitochondrial (putative) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_024537.3 | 1394 | Missense Mutation | AAC,GAC | N,D 527 | NP_078813.1 | |
XM_006719953.2 | 1394 | Missense Mutation | AAC,GAC | N,D 414 | XP_006720016.1 | |
XM_011521116.1 | 1394 | Missense Mutation | AAC,GAC | N,D 412 | XP_011519418.1 | |
XM_011521117.2 | 1394 | Intron | XP_011519419.1 | |||
XM_011521118.2 | 1394 | Intron | XP_011519420.1 | |||
XM_017020741.1 | 1394 | Missense Mutation | AAC,GAC | N,D 414 | XP_016876230.1 | |
XM_017020742.1 | 1394 | Intron | XP_016876231.1 |
NAXD - NAD(P)HX dehydratase | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |