Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCCAATTCCTGTGCCTCCTCCATGA[A/C]CTGTCTCTCCTCCTTTGGCTTTACT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616731 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC107984562 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LOC107984562 - serine/threonine-protein kinase Nek5-like | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
NEK5 - NIMA related kinase 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_199289.2 | Intron | NP_954983.1 | ||||
XM_006719808.3 | Intron | XP_006719871.2 | ||||
XM_011535066.2 | Intron | XP_011533368.1 | ||||
XM_011535068.2 | Intron | XP_011533370.1 | ||||
XM_017020555.1 | Intron | XP_016876044.1 | ||||
XM_017020556.1 | Intron | XP_016876045.1 | ||||
XM_017020557.1 | Intron | XP_016876046.1 |