Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGTAGCGACCCTCTTCAAGCTCCC[A/G]TTCAATCTCCCACCCGAGCTTGTAA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 602654 | ||||||||||||||||||||
Literature Links: |
FARP1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
FARP1 - FERM, ARH/RhoGEF and pleckstrin domain protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001001715.3 | 713 | Intron | NP_001001715.2 | |||
NM_001286839.1 | 713 | Intron | NP_001273768.1 | |||
NM_005766.3 | 713 | Intron | NP_005757.1 | |||
XM_011521046.2 | 713 | Intron | XP_011519348.1 | |||
XM_017020312.1 | 713 | Intron | XP_016875801.1 | |||
XM_017020313.1 | 713 | Intron | XP_016875802.1 |
RNF113B - ring finger protein 113B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_178861.4 | 713 | Missense Mutation | CGG,TGG | R,W 228 | NP_849192.1 |