Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTTAGGGCTACGCTTGGCCCTCCGG[T/C]GTCGGGTCAAGGCATGGCCCCGGCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 112262 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
BMP4 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
BMP4 - bone morphogenetic protein 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001202.4 | 1085 | Missense Mutation | CAC,CGC | H,R 287 | NP_001193.2 | |
NM_130850.3 | 1085 | Missense Mutation | CAC,CGC | H,R 287 | NP_570911.2 | |
XM_005268015.4 | 1085 | Missense Mutation | CAC,CGC | H,R 287 | XP_005268072.1 | |
XM_017021603.1 | 1085 | Missense Mutation | CAC,CGC | H,R 287 | XP_016877092.1 | |
XM_017021604.1 | 1085 | Missense Mutation | CAC,CGC | H,R 224 | XP_016877093.1 | |
XM_017021605.1 | 1085 | Missense Mutation | CAC,CGC | H,R 224 | XP_016877094.1 |
MIR5580 - microRNA 5580 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |