Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCCCAGAGGGGCCCCGAGATACATC[C/T]CCCAACTCTGAGACTTCCACATCTA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 614716 MIM: 615195 | ||||||||||||||||||||
Literature Links: |
CARMIL3 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CARMIL3 - capping protein regulator and myosin 1 linker 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_138360.3 | 134 | Silent Mutation | TCC,TCT | S,S 139 | NP_612369.3 | |
XM_011537300.2 | 134 | Silent Mutation | TCC,TCT | S,S 42 | XP_011535602.1 | |
XM_017021751.1 | 134 | Silent Mutation | TCC,TCT | S,S 19 | XP_016877240.1 |
DHRS4L1 - dehydrogenase/reductase 4 like 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |