Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAGTATTCTATGCTCACCTGGAAGA[C/T]GGGTACTACTTGGGATATCCCATGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603930 MIM: 600154 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
GPHN PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PIGH - phosphatidylinositol glycan anchor biosynthesis class H | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004569.3 | 562 | Missense Mutation | ATC,GTC | I,V 156 | NP_004560.1 | |
XM_011536838.2 | 562 | Missense Mutation | ATC,GTC | I,V 156 | XP_011535140.1 | |
XM_017021371.1 | 562 | Missense Mutation | ATC,GTC | I,V 155 | XP_016876860.1 | |
XM_017021372.1 | 562 | Missense Mutation | ATC,GTC | I,V 155 | XP_016876861.1 |
PLEKHH1 - pleckstrin homology, MyTH4 and FERM domain containing H1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |