Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CACAGTAATTGGCCTGAGACCGCTA[A/G]TGCTTCCTAAGAGGTATTTCCTAAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609398 MIM: 603930 MIM: 606958 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ATP6V1D PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ATP6V1D - ATPase H+ transporting V1 subunit D | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_015994.3 | 1368 | UTR 3 | NP_057078.1 |
LOC105370541 - uncharacterized LOC105370541 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MPP5 - membrane palmitoylated protein 5 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |